This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
ANTXR2
- Gene Name:
-
anthrax toxin receptor 2
- Gene Aliases:
-
CMG-2, CMG2, HFS, ISH, JHF
- Chromosome Location:
-
Chr.4: 79901617 - 80073472 on Build GRCh38
- UniGene ID:
-
Hs.162963
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
ANTXR2 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001145794.1 |
NP_001139266.1 |
8, 9
|
1469
|
|
| NM_001286780.1 |
NP_001273709.1 |
8, 9
|
632
|
|
| NM_001286781.1 |
NP_001273710.1 |
8, 9
|
1169
|
|
| NM_058172.5 |
NP_477520.2 |
8, 9
|
1469
|
|
| XM_011531587.2 |
XP_011529889.1 |
8, 9
|
632
|
|
| XM_017007711.1 |
XP_016863200.1 |
8, 9
|
1169
|
|
| GenBank mRNA |
AB209913.1 |
- |
8, 9
|
1057
|
|
| AK055636.1 |
- |
7, 8
|
520
|
|
| AL832851.1 |
- |
4, 5
|
300
|
|
| AY233452.1 |
- |
8, 9
|
709
|
|
| BC107876.1 |
- |
8, 9
|
635
|
6108432 |
| BC114374.1 |
- |
8, 9
|
581
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System