Search
Search

NIPA1| Gene Symbol: NIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142275.1 | NP_001135747.1 | 5 | 812 | ||
| NM_144599.4 | NP_653200.2 | 5 | 991 | |||
| GenBank mRNA | AB089319.1 | - | 1 | 490 | ||
| AK314073.1 | - | 5 | 952 | |||
| BC025678.1 | - | 5 | 812 | |||
| BC103705.1 | - | 6 | 938 | |||
| BX648722.1 | - | 1 | 450 | |||