Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256440.1 | NP_001243369.1 | 9 | 1056 | ||
NM_001256441.1 | NP_001243370.1 | 9 | 1051 | |||
NR_046202.1 | 9 | 1104 | ||||
XM_005258520.2 | XP_005258577.1 | 9 | 1045 | |||
XM_011526460.2 | XP_011524762.1 | 9 | 1044 | |||
XM_017026291.1 | XP_016881780.1 | 9 | 1056 | |||
XM_017026292.1 | XP_016881781.1 | 8 | 965 | |||
XM_017026293.1 | XP_016881782.1 | 9 | 1056 | |||
XM_017026294.1 | XP_016881783.1 | 6 | 774 | |||
XM_017026295.1 | XP_016881784.1 | 10 | 1154 | |||
GenBank mRNA | AI394044.1 | - | 2 | 221 | ||
AK094207.1 | - | 9 | 1044 | |||
AL834131.1 | - | 9 | 1081 | |||
BC023623.2 | - | 9 | 1082 | 4662735 | ||
BC027935.1 | - | 9 | 1072 | 5202287 | ||
CR936760.1 | - | 9 | 1057 |