Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS134233
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
7
861
NM_152385.2
NP_689598.2
9
1312
XM_005264136.2
XP_005264193.1
8
1180
XM_006711937.3
XP_006712000.2
6
619
XM_006711938.3
XP_006712001.1
3
239
XM_006711939.3
XP_006712002.1
2
111
XM_011532519.2
XP_011530821.1
9
4205
XM_011532520.2
XP_011530822.1
9
1483
XM_011532521.2
XP_011530823.1
10
1639
XM_011532522.2
XP_011530824.1
10
1626
XM_011532523.2
XP_011530825.1
9
1715
XM_011532524.1
XP_011530826.1
8
1097
XM_011532525.2
XP_011530827.1
8
1163
XM_011532527.2
XP_011530829.1
9
4807
XM_011532528.1
XP_011530830.1
8
1707
XM_011532529.2
XP_011530831.1
7
1666
XM_011532530.1
XP_011530832.1
7
1427
XM_011532534.1
XP_011530836.1
7
1046
XM_011532535.2
XP_011530837.1
5
556
XM_017003333.1
XP_016858822.1
7
4007
XM_017003334.1
XP_016858823.1
7
1285
XM_017003335.1
XP_016858824.1
7
4375
XM_017003336.1
XP_016858825.1
8
1796
XM_017003337.1
XP_016858826.1
6
885
XM_017003338.1
XP_016858827.1
6
863
XM_017003339.1
XP_016858828.1
9
4205
XM_017003340.1
XP_016858829.1
7
8307
XM_017003341.1
XP_016858830.1
7
1792
XM_017003342.1
XP_016858831.1
6
1293
XM_017003343.1
XP_016858832.1
6
1509
XM_017003344.1
XP_016858833.1
6
1597
XM_017003345.1
XP_016858834.1
5
1467
XM_017003346.1
XP_016858835.1
7
4007
XM_017003347.1
XP_016858836.1
4
1335
XM_017003348.1
XP_016858837.1
9
4205
XM_017003349.1
XP_016858838.1
4
3754
XM_017003350.1
XP_016858839.1
4
1265
GenBank mRNA
AK315683.1
-
9
1270
BC029502.1
-
9
1312
5270720
BC036908.1
-
7
861
5166064
BM046729.1
-
2
94
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0