Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLHC1 SIRNA Assays ›
          Gene Symbol:
          CLHC1
          Gene Name:
          clathrin heavy chain linker domain containing 1
          Gene Aliases:
          C2orf63
          Chromosome Location:
          Chr.2: 55172547 - 55232563 on Build GRCh38
          UniGene ID:
          Hs.347014
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS134234
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLHC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135598.1 NP_001129070.1 9, 10 1287
          NM_152385.2 NP_689598.2 11, 12 1738
          XM_005264136.2 XP_005264193.1 10, 11 1606
          XM_006711937.3 XP_006712000.2 8, 9 1045
          XM_006711938.3 XP_006712001.1 5, 6 665
          XM_006711939.3 XP_006712002.1 4, 5 537
          XM_011532519.2 XP_011530821.1 11, 12 4631
          XM_011532520.2 XP_011530822.1 11, 12 1909
          XM_011532521.2 XP_011530823.1 12, 13 2065
          XM_011532522.2 XP_011530824.1 12, 13 2052
          XM_011532523.2 XP_011530825.1 11, 12 2141
          XM_011532524.1 XP_011530826.1 10, 11 1523
          XM_011532525.2 XP_011530827.1 10, 11 1589
          XM_011532527.2 XP_011530829.1 11, 12 5233
          XM_011532528.1 XP_011530830.1 10, 11 2133
          XM_011532529.2 XP_011530831.1 9, 10 2092
          XM_011532530.1 XP_011530832.1 9, 10 1853
          XM_011532534.1 XP_011530836.1 9, 10 1472
          XM_011532535.2 XP_011530837.1 7, 8 982
          XM_017003333.1 XP_016858822.1 9, 10 4433
          XM_017003334.1 XP_016858823.1 9, 10 1711
          XM_017003335.1 XP_016858824.1 9, 10 4801
          XM_017003336.1 XP_016858825.1 10, 11 2222
          XM_017003337.1 XP_016858826.1 8, 9 1311
          XM_017003338.1 XP_016858827.1 8, 9 1289
          XM_017003340.1 XP_016858829.1 9, 10 8733
          XM_017003341.1 XP_016858830.1 9, 10 2218
          XM_017003342.1 XP_016858831.1 8, 9 1719
          XM_017003343.1 XP_016858832.1 8, 9 1935
          XM_017003344.1 XP_016858833.1 8, 9 2023
          XM_017003345.1 XP_016858834.1 7, 8 1893
          XM_017003347.1 XP_016858836.1 6, 7 1761
          XM_017003349.1 XP_016858838.1 6, 7 4180
          XM_017003350.1 XP_016858839.1 6, 7 1691
          GenBank mRNA AK056000.1 - 4, 5 684
          AK315683.1 - 11, 12 1696
          BC029502.1 - 11, 12 1738 5270720
          BC036908.1 - 9, 10 1287 5166064
          BM046729.1 - 5, 6 645
          DA249479.1 - 1, 2 33

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hqkpn:80/100.66.73.1:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0