Search
Search

C7orf31| Gene Symbol: C7orf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138811.3 | NP_620166.3 | 7 | 1184 | ||
| XM_005249619.3 | XP_005249676.1 | 7 | 1033 | |||
| XM_005249620.3 | XP_005249677.1 | 8 | 1564 | |||
| XM_011515125.2 | XP_011513427.1 | 3 | 75 | |||
| GenBank mRNA | AK097248.1 | - | 8 | 905 | ||
| AK098189.1 | - | 2 | 95 | |||
| BC043269.2 | - | 7 | 1287 | 5297014 | ||
| BC060879.1 | - | 7 | 1097 | 5298874 | ||
| BX640851.1 | - | 4 | 254 | |||