Search
Search

ZNF664| Gene Symbol: ZNF664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204298.1 | NP_001191227.1 | 5 | 1450 | ||
| NM_152437.2 | NP_689650.1 | 5 | 1502 | |||
| GenBank mRNA | AK056034.1 | - | 5 | 1409 | ||
| AL834266.1 | - | 6 | 2303 | |||
| BC017960.1 | - | 1 | 766 | 4608465 | ||
| BC051696.1 | - | 5 | 1432 | 5276234 | ||
| BC093848.1 | - | 1 | 513 | 7939693 | ||
| BC101530.1 | - | 1 | 513 | 8069036 | ||
| BX647762.1 | - | 6 | 1422 | |||
| L41669.1 | - | 1 | 406 | |||