Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CMTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178818.2 | NP_848933.1 | 2 | 384 | ||
NM_181521.2 | NP_852662.1 | 2 | 384 | |||
XM_011522882.1 | XP_011521184.1 | 2 | 292 | |||
XM_017022954.1 | XP_016878443.1 | 2 | 288 | |||
GenBank mRNA | AF479814.1 | - | 2 | 384 | ||
AF521889.1 | - | 2 | 384 | |||
AF521890.1 | - | 2 | 297 | |||
BC093679.1 | - | 2 | 266 | 7939524 |