Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039887.2 | NP_001034976.2 | 3 | 318 | ||
XM_011526528.1 | XP_011524830.1 | 3 | 308 | |||
XM_011526529.2 | XP_011524831.1 | 3 | 322 | |||
XM_011526530.1 | XP_011524832.1 | 3 | 308 | |||
XM_011526531.1 | XP_011524833.1 | 3 | 308 | |||
XM_011526532.1 | XP_011524834.1 | 3 | 308 | |||
XM_011526533.2 | XP_011524835.1 | 3 | 308 | |||
XM_011526534.1 | XP_011524836.1 | 3 | 308 | |||
XM_011526536.2 | XP_011524838.1 | 3 | 322 | |||
GenBank mRNA | AK055219.1 | - | 3 | 277 | ||
AK124144.1 | - | 1 | 1727 | |||
AL833894.1 | - | 3 | 282 | |||
BC017947.1 | - | 3 | 267 | |||
BC110893.1 | - | 2 | 249 | 3914279 | ||
DB086668.1 | - | 3 | 310 |