Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL23R SIRNA Assays ›
          Gene Symbol:
          IL23R
          Gene Name:
          interleukin 23 receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 67138638 - 67259979 on Build GRCh38
          UniGene ID:
          Hs.677426
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS135534
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL23R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144701.2 NP_653302.2 5 605
          XM_011540790.2 XP_011539092.1 5 1073
          XM_011540791.2 XP_011539093.1 5 590
          GenBank mRNA AF461422.1 - 5 605
          AM990313.1 - 2 291
          AM990315.1 - 2 82
          AM990316.1 - 2 82
          AM990317.1 - 2 82
          AM990318.1 - 2 82
          AM990320.1 - 2 82
          AM990321.1 - 2 82
          AM990323.1 - 2 77
          AM990325.1 - 2 82
          AM990326.1 - 2 82
          AM990331.1 - 2 82
          AM990332.1 - 2 82
          AM990333.1 - 2 77
          AM990335.1 - 2 82
          AM990336.1 - 2 77
          AY937250.1 - 3 412
          AY937253.1 - 3 412
          AY937254.1 - 3 412
          AY937255.1 - 3 412
          FM212237.1 - 3 241
          FM212239.1 - 3 241
          FM212241.1 - 3 241

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          positive regulation of T-helper 1 type immune response
          inflammatory response
          positive regulation of activation of JAK2 kinase activity
          response to lipopolysaccharide
          negative regulation of interleukin-10 production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of natural killer cell proliferation
          response to interferon-gamma
          interleukin-23-mediated signaling pathway
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat4 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          positive regulation of memory T cell differentiation
          positive regulation of osteoclast differentiation
          defense response to Gram-negative bacterium
          positive regulation of NK T cell activation
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          interleukin-12 receptor binding
          protein binding
          interleukin-23 binding
          interleukin-23 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0