Search
Search

LRRC71| Gene Symbol: LRRC71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144702.2 | NP_653303.2 | 7 | 849 | ||
| XM_005244926.3 | XP_005244983.1 | 7 | 873 | |||
| XM_005244927.3 | XP_005244984.1 | 7 | 873 | |||
| XM_005244928.2 | XP_005244985.1 | 7 | 873 | |||
| XM_006711185.3 | XP_006711248.1 | 8 | 663 | |||
| XM_006711186.3 | XP_006711249.1 | 7 | 873 | |||
| XM_006711187.3 | XP_006711250.1 | 7 | 873 | |||
| XM_011509239.2 | XP_011507541.1 | 3 | 345 | |||
| XM_011509240.2 | XP_011507542.1 | 3 | 327 | |||
| XM_011509241.2 | XP_011507543.1 | 7 | 873 | |||
| XM_017000459.1 | XP_016855948.1 | 7 | 873 | |||
| XM_017000460.1 | XP_016855949.1 | 7 | 1181 | |||
| XM_017000461.1 | XP_016855950.1 | 8 | 719 | |||
| XM_017000462.1 | XP_016855951.1 | 7 | 622 | |||
| GenBank mRNA | AK057446.1 | - | 8 | 678 | ||
| AK292342.1 | - | 7 | 849 | |||
| BC033790.1 | - | 7 | 831 | 5163973 | ||