Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 10 | 3115 | ||
NM_178835.4 | NP_849157.2 | 10 | 3115 | |||
XM_011531634.2 | XP_011529936.1 | 10 | 3162 | |||
XM_011531635.2 | XP_011529937.1 | 11 | 8649 | |||
XM_011531645.2 | XP_011529947.1 | 6 | 900 | |||
XM_017007768.1 | XP_016863257.1 | 10 | 3157 | |||
XM_017007769.1 | XP_016863258.1 | 10 | 9408 | |||
XM_017007770.1 | XP_016863259.1 | 9 | 2929 | |||
XM_017007771.1 | XP_016863260.1 | 10 | 3161 | |||
XM_017007772.1 | XP_016863261.1 | 10 | 3161 | |||
XM_017007773.1 | XP_016863262.1 | 10 | 3161 | |||
XM_017007774.1 | XP_016863263.1 | 10 | 3161 | |||
XM_017007775.1 | XP_016863264.1 | 10 | 3161 | |||
XM_017007777.1 | XP_016863266.1 | 7 | 899 | |||
GenBank mRNA | AB074279.1 | - | 3 | 235 | ||
AK091130.1 | - | 10 | 2585 | |||
AK094136.1 | - | 5 | 404 | |||
AK225805.1 | - | 10 | 2790 | |||
BC117407.1 | - | 10 | 2626 | 40125958 | ||
BC143577.1 | - | 10 | 2626 | 9052088 |