Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC38A9 SIRNA Assays ›
Gene Symbol:
SLC38A9
Gene Name:
solute carrier family 38 member 9
Gene Aliases:
URLC11
Chromosome Location:
Chr.5: 55625845 - 55712344 on Build GRCh38
UniGene ID:
Hs.742101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS135862
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC38A9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258286.1
NP_001245215.1
2
545
NM_001258287.1
NP_001245216.1
3
649
NM_173514.3
NP_775785.2
4
395
NR_047649.1
2
545
XM_006714537.2
XP_006714600.1
6
629
XM_006714538.3
XP_006714601.1
5
516
XM_006714539.3
XP_006714602.1
5
559
XM_011543173.1
XP_011541475.1
6
617
XM_011543174.1
XP_011541476.1
7
715
XM_011543176.1
XP_011541478.1
6
629
XM_011543177.1
XP_011541479.1
6
629
XM_011543178.1
XP_011541480.1
6
629
XM_011543180.1
XP_011541482.1
6
629
XM_011543182.1
XP_011541484.1
6
629
XM_011543183.1
XP_011541485.1
6
629
XM_011543184.1
XP_011541486.1
6
629
XM_017009077.1
XP_016864566.1
4
327
XM_017009078.1
XP_016864567.1
4
404
XM_017009079.1
XP_016864568.1
4
327
XM_017009080.1
XP_016864569.1
5
516
XM_017009081.1
XP_016864570.1
4
327
XM_017009082.1
XP_016864571.1
4
327
GenBank mRNA
AB105188.1
-
4
327
AK075190.1
-
4
327
AK127988.1
-
3
289
AK301850.1
-
2
207
BC066891.1
-
4
384
4373965
BC101362.2
-
4
326
40025719
DC371481.1
-
4
395
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transmembrane transport
amino acid transport
positive regulation of TOR signaling
cellular response to amino acid stimulus
protein binding
amino acid transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5trjz:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0