Search
Search

BTNL9| Gene Symbol: BTNL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308245.1 | NP_001295174.1 | 4 | 824 | ||
| NM_152547.4 | NP_689760.2 | 4 | 824 | |||
| XM_006714823.3 | XP_006714886.1 | 4 | 820 | |||
| XM_006714824.3 | XP_006714887.1 | 4 | 820 | |||
| XM_006714825.3 | XP_006714888.1 | 5 | 1211 | |||
| XM_011534444.2 | XP_011532746.1 | 4 | 820 | |||
| XM_017009113.1 | XP_016864602.1 | 4 | 820 | |||
| XM_017009114.1 | XP_016864603.1 | 4 | 691 | |||
| XM_017009115.1 | XP_016864604.1 | 4 | 820 | |||
| GenBank mRNA | AK057097.1 | - | 4 | 812 | ||
| AK298113.1 | - | 3 | 651 | |||
| AY358358.1 | - | 4 | 810 | |||
| BC062459.1 | - | 4 | 683 | 6185476 | ||