Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARMCX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256155.2 | NP_001243084.2 | 2 | 809 | ||
NR_028407.2 | 9 | 1429 | ||||
NR_045861.1 | 6 | 1320 | ||||
NR_045864.1 | 6 | 1362 | ||||
GenBank mRNA | AK096955.1 | - | 6 | 1118 | ||
AK126987.1 | - | 6 | 1183 | |||
AK292543.1 | - | 6 | 1153 | |||
BC032236.1 | - | 6 | 1099 | |||
CR933662.1 | - | 9 | 1404 |