Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFPM1 SIRNA Assays ›
          Gene Symbol:
          ZFPM1
          Gene Name:
          zinc finger protein, FOG family member 1
          Gene Aliases:
          FOG, FOG1, ZC2HC11A, ZNF408, ZNF89A
          Chromosome Location:
          Chr.16: 88451782 - 88535166 on Build GRCh38
          UniGene ID:
          Hs.632218
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS136279
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFPM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153813.2 NP_722520.2 6, 7 733
          XM_011522912.2 XP_011521214.1 6, 7 4582
          XM_011522914.2 XP_011521216.1 6, 7 817
          XM_011522917.2 XP_011521219.1 4, 5 643
          XM_017022982.1 XP_016878471.1 4, 5 522
          GenBank mRNA AF488691.1 - 6, 7 1022

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          T-helper cell lineage commitment
          outflow tract morphogenesis
          atrioventricular valve morphogenesis
          mitral valve formation
          tricuspid valve formation
          transcription, DNA-templated
          heart development
          blood coagulation
          regulation of definitive erythrocyte differentiation
          erythrocyte differentiation
          megakaryocyte differentiation
          platelet formation
          granulocyte differentiation
          negative regulation of protein binding
          regulation of chemokine production
          embryonic hemopoiesis
          megakaryocyte development
          positive regulation of interferon-gamma biosynthetic process
          negative regulation of interleukin-4 biosynthetic process
          negative regulation of fat cell differentiation
          cardiac muscle tissue morphogenesis
          definitive erythrocyte differentiation
          primitive erythrocyte differentiation
          negative regulation of mast cell differentiation
          ventricular septum morphogenesis
          atrial septum morphogenesis
          transcriptional activation by promoter-enhancer looping
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II activating transcription factor binding
          protein binding
          transcription factor binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0