Search
Search

ZNF564| Gene Symbol: ZNF564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144976.3 | NP_659413.1 | 4 | 1850 | ||
| GenBank mRNA | AK092760.1 | - | 4 | 1811 | ||
| AL834146.1 | - | 4 | 5110 | |||
| BC028367.1 | - | 4 | 1787 | |||
| BC065023.1 | - | 4 | 1760 | 6044821 | ||
| BC136728.1 | - | 4 | 1686 | 9020718 | ||
| BC136729.1 | - | 4 | 1686 | 9020719 | ||
| BX647959.1 | - | 4 | 1822 | |||