Search
Search

KANK4| Gene Symbol: KANK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320269.1 | NP_001307198.1 | 8 | 1230 | ||
| NM_181712.4 | NP_859063.3 | 9 | 3112 | |||
| XM_011540843.2 | XP_011539145.1 | 9 | 2997 | |||
| XM_011540845.2 | XP_011539147.1 | 9 | 2991 | |||
| XM_017000484.1 | XP_016855973.1 | 9 | 1366 | |||
| XM_017000485.1 | XP_016855974.1 | 7 | 1298 | |||
| GenBank mRNA | AB284127.1 | - | 9 | 3112 | ||
| AK091941.1 | - | 8 | 1117 | |||
| AK096259.1 | - | 3 | 496 | |||
| AK127461.1 | - | 6 | 2038 | |||
| AL832644.2 | - | 7 | 1864 | |||
| BC041418.1 | - | 7 | 1332 | |||
| BC060866.1 | - | 9 | 3112 | 30344114 | ||