Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168316.1 | NP_001161788.1 | 5 | 1284 | ||
NM_174914.3 | NP_777574.2 | 6 | 1386 | |||
NR_031764.1 | 5 | 854 | ||||
XM_011513988.2 | XP_011512290.1 | 7 | 1428 | |||
XM_017009150.1 | XP_016864639.1 | 6 | 1437 | |||
GenBank mRNA | AK075383.1 | - | 6 | 1347 | ||
AK300751.1 | - | 5 | 1245 | |||
AK303154.1 | - | 5 | 815 | |||
AY358416.1 | - | 6 | 1321 | |||
BC103924.1 | - | 6 | 1335 | 40007344 | ||
BC103925.1 | - | 6 | 1335 | 40007348 |