Search
Search

GDPD4| Gene Symbol: GDPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182833.1 | NP_878253.1 | 10 | 987 | ||
| XM_011544826.1 | XP_011543128.1 | 10 | 2718 | |||
| XM_011544833.1 | XP_011543135.1 | 10 | 2718 | |||
| XM_011544834.1 | XP_011543136.1 | 11 | 1213 | |||
| XM_017017348.1 | XP_016872837.1 | 11 | 1157 | |||
| XM_017017349.1 | XP_016872838.1 | 10 | 1074 | |||
| XM_017017350.1 | XP_016872839.1 | 10 | 2589 | |||
| XM_017017351.1 | XP_016872840.1 | 10 | 945 | |||
| GenBank mRNA | AY313782.1 | - | 10 | 987 | ||
| AY326450.1 | - | 10 | 987 | |||