Search Thermo Fisher Scientific
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175061.3 | NP_778231.2 | 3 | 475 | ||
XM_006715656.1 | XP_006715719.1 | 4 | 351 | |||
XM_017011831.1 | XP_016867320.1 | 3 | 243 | |||
XM_017011832.1 | XP_016867321.1 | 3 | 175 | |||
GenBank mRNA | AK091311.1 | - | 3 | 400 | ||
AL832491.1 | - | 1 | 53 | |||
AL834234.1 | - | 2 | 194 | |||
AY372319.1 | - | 3 | 253 | |||
BC042441.1 | - | 3 | 263 | 4831063 | ||
BC047229.1 | - | 3 | 262 | |||
DA864624.1 | - | 3 | 475 |