Search
Search

SCUBE3| Gene Symbol: SCUBE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152753.3 | NP_689966.2 | 4, 5 | 828 | ||
| XM_005248943.1 | XP_005249000.1 | 4, 5 | 843 | |||
| XM_005248947.1 | XP_005249004.1 | 4, 5 | 843 | |||
| XM_011514404.2 | XP_011512706.1 | 4, 5 | 543 | |||
| GenBank mRNA | AF452494.1 | - | 4, 5 | 477 | ||
| AK092062.1 | - | 3, 4 | 339 | |||
| AK291218.1 | - | 5, 6 | 549 | |||
| AY639608.1 | - | 4, 5 | 471 | |||