Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCO4 SIRNA Assays ›
Gene Symbol:
TMCO4
Gene Name:
transmembrane and coiled-coil domains 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 19682213 - 19800385 on Build GRCh38
UniGene ID:
Hs.656313
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS137891
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCO4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181719.4
NP_859070.3
16
1945
XM_005245822.4
XP_005245879.1
16
1961
XM_005245824.4
XP_005245881.1
16
2025
XM_005245825.4
XP_005245882.1
15
1826
XM_005245827.3
XP_005245884.1
15
1841
XM_006710551.3
XP_006710614.1
16
2050
XM_011541179.2
XP_011539481.1
19
2420
XM_011541180.2
XP_011539482.1
19
2575
XM_011541181.1
XP_011539483.1
18
2249
XM_011541185.2
XP_011539487.1
16
1968
XM_011541186.2
XP_011539488.1
16
2065
XM_017000913.1
XP_016856402.1
15
1849
XM_017000914.1
XP_016856403.1
18
2469
XM_017000915.1
XP_016856404.1
18
2191
XM_017000916.1
XP_016856405.1
16
2103
XM_017000917.1
XP_016856406.1
17
2318
XM_017000918.1
XP_016856407.1
18
2515
XM_017000919.1
XP_016856408.1
17
2242
XM_017000920.1
XP_016856409.1
16
2460
XM_017000921.1
XP_016856410.1
17
2267
XM_017000922.1
XP_016856411.1
15
1926
GenBank mRNA
AK127543.1
-
5
722
AL832390.1
-
4
456
BC053600.1
-
14
1804
5744211
BX640987.1
-
16
1946
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0