Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPNT SIRNA Assays ›
          Gene Symbol:
          NPNT
          Gene Name:
          nephronectin
          Gene Aliases:
          EGFL6L, POEM
          Chromosome Location:
          Chr.4: 105895440 - 105971671 on Build GRCh38
          UniGene ID:
          Hs.518921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS137946
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPNT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033047.2 NP_001028219.1 7 956
          NM_001184690.1 NP_001171619.1 8 1007
          NM_001184691.1 NP_001171620.1 8 1046
          NM_001184692.1 NP_001171621.1 7 956
          NM_001184693.1 NP_001171622.1 8 1046
          XM_005262888.4 XP_005262945.1 9 1084
          XM_005262890.1 XP_005262947.1 7 731
          XM_011531820.2 XP_011530122.1 9 1087
          XM_011531822.2 XP_011530124.1 8 1036
          XM_011531823.2 XP_011530125.1 9 1087
          XM_011531824.2 XP_011530126.1 8 997
          XM_011531825.2 XP_011530127.1 7 946
          XM_017007984.1 XP_016863473.1 9 1087
          GenBank mRNA AK290029.1 - 7 903
          AK295772.1 - 8 1007
          AK302477.1 - 8 1045
          AK304279.1 - 7 955
          AK304721.1 - 8 993
          AL832465.1 - 7 870
          AW607534.1 - 2 253
          AY358336.1 - 7 945
          BC131587.1 - 7 803

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          branching involved in ureteric bud morphogenesis
          cell-matrix adhesion
          positive regulation of alkaline phosphatase activity
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          positive regulation of transforming growth factor beta receptor signaling pathway
          cell-cell adhesion mediated by integrin
          establishment of protein localization
          positive regulation of osteoblast differentiation
          positive regulation of smooth muscle contraction
          positive regulation of ERK1 and ERK2 cascade
          cellular response to tumor necrosis factor
          pilomotor reflex
          positive regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          integrin binding
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0