Search Thermo Fisher Scientific
Gene Symbol: ZAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175619.2 | NP_783318.1 | 1, 2 | 1006 | ||
GenBank mRNA | AY191416.1 | - | 1, 2 | 966 | ||
BC136265.1 | - | 1, 2 | 1006 | 9020252 |