Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C11orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198498.2 | NP_940900.1 | 4 | 839 | ||
XM_011542803.2 | XP_011541105.1 | 5 | 982 | |||
XM_011542804.2 | XP_011541106.1 | 4 | 765 | |||
GenBank mRNA | AI864880.1 | - | 1 | 371 | ||
BC039669.1 | - | 4 | 839 | 5167284 | ||
HY297763.1 | - | 1 | 391 |