Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANKRD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130015.1 | NP_001123487.1 | 1 | 198 | ||
NM_001304459.1 | NP_001291388.1 | 1 | 198 | |||
NM_001304460.1 | NP_001291389.1 | 1 | 198 | |||
NM_182608.3 | NP_872414.3 | 1 | 198 | |||
XM_017019258.1 | XP_016874747.1 | 1 | 91 | |||
GenBank mRNA | AF517108.1 | - | 1 | 97 | ||
AF517109.1 | - | 1 | 97 | |||
AK125517.1 | - | 1 | 247 | |||
BC121152.1 | - | 1 | 198 | |||
BC121153.2 | - | 1 | 195 | 40121303 |