Search
Search

VHLL| Gene Symbol: VHLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004319.2 | NP_001004319.1 | 1 | 614 | ||
| GenBank mRNA | AY494836.1 | - | 1 | 276 | ||
| BC130596.1 | - | 1 | 241 | 40146627 | ||
| BC130598.1 | - | 1 | 241 | 40146629 | ||