Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2B SIRNA Assays ›
Gene Symbol:
CAMK2B
Gene Name:
calcium/calmodulin dependent protein kinase II beta
Gene Aliases:
CAM2, CAMK2, CAMKB
Chromosome Location:
Chr.7: 44217150 - 44325994 on Build GRCh38
UniGene ID:
Hs.351887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS141028
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220.4
NP_001211.3
3
387
NM_001293170.1
NP_001280099.1
3
387
NM_172078.2
NP_742075.1
3
387
NM_172079.2
NP_742076.1
3
387
NM_172080.2
NP_742077.1
3
387
NM_172081.2
NP_742078.1
3
387
NM_172082.2
NP_742079.1
3
387
NM_172083.2
NP_742080.1
3
387
NM_172084.2
NP_742081.1
3
387
XM_005249862.3
XP_005249919.1
3
263
XM_005249864.3
XP_005249921.1
3
263
XM_006715781.2
XP_006715844.1
3
263
XM_006715784.2
XP_006715847.1
3
263
XM_011515547.1
XP_011513849.1
3
263
XM_011515549.1
XP_011513851.1
3
263
XM_011515550.1
XP_011513852.1
3
263
XM_011515551.1
XP_011513853.1
3
263
XM_011515552.1
XP_011513854.1
3
263
XM_011515553.1
XP_011513855.1
3
263
XM_011515554.1
XP_011513856.1
3
263
XM_011515555.1
XP_011513857.1
3
263
XM_011515556.1
XP_011513858.1
3
263
XM_011515557.1
XP_011513859.1
3
263
XM_011515558.2
XP_011513860.1
3
263
XM_011515559.1
XP_011513861.1
3
172
XM_017012660.1
XP_016868149.1
3
263
XM_017012661.1
XP_016868150.1
3
263
XM_017012662.1
XP_016868151.1
3
263
XM_017012663.1
XP_016868152.1
3
172
XM_017012664.1
XP_016868153.1
3
263
XM_017012665.1
XP_016868154.1
3
263
XM_017012666.1
XP_016868155.1
3
172
XM_017012667.1
XP_016868156.1
3
172
XM_017012668.1
XP_016868157.1
3
172
XM_017012669.1
XP_016868158.1
3
172
XM_017012670.1
XP_016868159.1
3
172
GenBank mRNA
AF078803.1
-
3
180
AF081572.1
-
3
180
AF081924.1
-
3
180
AF083419.1
-
3
180
AF112471.1
-
3
223
AF112472.1
-
3
223
AF140350.1
-
3
180
AJ252236.1
-
3
264
AJ252237.1
-
3
187
AJ252238.1
-
3
177
AK222703.1
-
3
211
AK290148.1
-
3
252
AK294138.1
-
3
231
AK315663.1
-
3
294
BC019070.2
-
3
381
5014712
BI550751.1
-
3
392
BX445032.2
-
3
196
DA069361.1
-
2
39
DB175099.1
-
3
165
U23460.1
-
3
252
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
signal transduction
positive regulation of neuron projection development
regulation of skeletal muscle adaptation
positive regulation of phospholipase A2 activity
positive regulation of GTPase activity
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
regulation of synapse structural plasticity
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of dendritic spine development
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
regulation of cellular response to heat
actin binding
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein kinase binding
protein homodimerization activity
phospholipase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0