Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANAPC15 SIRNA Assays ›
          Gene Symbol:
          ANAPC15 LRTOMT
          Gene Name:
          anaphase promoting complex subunit 15
          leucine rich transmembrane and O-methyltransferase domain containing
          Gene Aliases:
          APC15, C11orf51, HSPC020
          CFAP111, DFNB63, LRRC51
          Chromosome Location:
          Chr.11: 72106254 - 72112793 on Build GRCh38
          Chromosome Location:
          Chr.11: 72079982 - 72110782 on Build GRCh38
          UniGene ID:
          Hs.38044
          Hs.317243
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS141119
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANAPC15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278485.1 NP_001265414.1 6 585
          NM_001278486.1 NP_001265415.1 6 580
          NM_001278487.1 NP_001265416.1 6 568
          NM_001278488.1 NP_001265417.1 6 484
          NM_001278489.1 NP_001265418.1 6 543
          NM_001278490.1 NP_001265419.1 5 488
          NM_001278491.1 NP_001265420.1 6 471
          NM_001278492.1 NP_001265421.1 6 498
          NM_001278493.1 NP_001265422.1 6 480
          NM_001278494.1 NP_001265423.1 6 755
          NM_014042.2 NP_054761.1 6 573
          XM_017017496.1 XP_016872985.1 6 599
          XM_017017497.1 XP_016872986.1 6 575
          GenBank mRNA AF077206.1 - 6 574
          AK222622.1 - 6 572
          AL080071.1 - 6 577
          BC005156.2 - 6 566 2964612
          BC005393.1 - 6 567 4051407
          BE616852.1 - 6 472
          BG752082.1 - 6 564
          BG764016.1 - 6 555
          BU532056.1 - 6 488
          BU856100.1 - 6 517
          BU930156.1 - 5 401
          CN306434.1 - 6 543
          HY080534.1 - 6 470
          Gene Symbol: LRTOMT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145308.4 NP_001138780.1 7 2719
          NM_001145309.3 NP_001138781.1 9 2940
          NM_001145310.3 NP_001138782.1 9 2820
          GenBank mRNA AF289588.1 - 5 1801
          EU627069.1 - 9 2412
          EU627070.1 - 9 2292

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001170435, XM_006229860, XM_006229869, XM_006229862, XM_008759705, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001291352, NM_001291348, NM_001291349, NM_001291353, NM_027532, and others.

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic nuclear division
          anaphase-promoting complex-dependent catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          regulation of mitotic cell cycle spindle assembly checkpoint
          sensory perception of sound
          methylation
          neurotransmitter catabolic process
          catecholamine catabolic process
          auditory receptor cell development
          catechol O-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0