Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK1 SIRNA Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
UniGene ID:
Hs.654438
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS141213
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000037.3
NP_000028.3
43
5982
NM_001142445.1
NP_001135917.1
4
926
NM_001142446.1
NP_001135918.1
43
6168
NM_020475.2
NP_065208.2
42
5847
NM_020476.2
NP_065209.2
42
5922
NM_020477.2
NP_065210.2
42
5436
NM_020478.4
NP_065211.2
5
986
NM_020480.4
NP_065213.2
3
785
XM_005273476.4
XP_005273533.1
44
6280
XM_011544490.2
XP_011542792.1
45
6187
XM_011544491.2
XP_011542793.1
46
6246
XM_011544494.2
XP_011542796.1
44
6141
XM_011544495.2
XP_011542797.1
45
6200
XM_011544496.2
XP_011542798.1
45
6112
XM_011544500.2
XP_011542802.1
42
6024
XM_011544501.2
XP_011542803.1
43
6084
XM_011544502.2
XP_011542804.1
42
5949
XM_011544503.2
XP_011542805.1
45
5713
XM_011544504.2
XP_011542806.1
42
5538
XM_011544505.2
XP_011542807.1
43
5598
XM_017013319.1
XP_016868808.1
44
6162
XM_017013320.1
XP_016868809.1
45
6182
XM_017013321.1
XP_016868810.1
45
6131
XM_017013322.1
XP_016868811.1
44
6093
XM_017013323.1
XP_016868812.1
45
6107
XM_017013324.1
XP_016868813.1
43
6220
XM_017013325.1
XP_016868814.1
44
6032
XM_017013326.1
XP_016868815.1
44
6040
XM_017013327.1
XP_016868816.1
45
5700
XM_017013328.1
XP_016868817.1
44
5655
XM_017013329.1
XP_016868818.1
44
5792
GenBank mRNA
AB209418.1
-
43
6168
AK126647.1
-
27
3376
AK129864.1
-
6
800
AK223578.1
-
5
907
BC007930.2
-
4
826
BC030957.1
-
5
974
M28880.1
-
43
5982
X16609.1
-
42
5922
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein targeting to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0