Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C1QA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015991.2 | NP_057075.1 | 2 | 147 | ||
GenBank mRNA | AF135157.1 | - | 1 | 74 | ||
AF260332.1 | - | 1 | 78 | |||
AK311980.1 | - | 2 | 90 | |||
BC030153.2 | - | 2 | 117 | 4850418 | ||
BC071986.1 | - | 1 | 83 | 6300836 | ||
BG548439.1 | - | 2 | 148 |