Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP8 SIRNA Assays ›
          Gene Symbol:
          CASP8
          Gene Name:
          caspase 8
          Gene Aliases:
          ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
          Chromosome Location:
          Chr.2: 201233443 - 201287711 on Build GRCh38
          UniGene ID:
          Hs.599762
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS141462
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASP8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080124.1 NP_001073593.1 4, 5 645
          NM_001080125.1 NP_001073594.1 3, 4 788
          NM_001228.4 NP_001219.2 5, 6 809
          NM_033355.3 NP_203519.1 4, 5 619
          NM_033356.3 NP_203520.1 3, 4 550
          NM_033358.3 NP_203522.1 4, 5 713
          NR_111983.1 3, 4 788
          XM_005246885.1 XP_005246942.1 3, 4 608
          XM_005246886.1 XP_005246943.1 3, 4 534
          XM_005246887.1 XP_005246944.1 4, 5 686
          XM_005246888.1 XP_005246945.1 4, 5 628
          XM_005246889.1 XP_005246946.1 3, 4 505
          XM_005246890.3 XP_005246947.1 3, 4 512
          XM_005246891.4 XP_005246948.1 3, 4 752
          XM_005246892.1 XP_005246949.1 3, 4 505
          XM_005246893.2 XP_005246950.1 3, 4 608
          XM_005246894.3 XP_005246951.1 4, 5 683
          XM_005246895.2 XP_005246952.1 3, 4 608
          XM_006712789.1 XP_006712852.1 4, 5 645
          XM_006712790.3 XP_006712853.1 3, 4 524
          XM_006712793.2 XP_006712856.1 4, 5 686
          GenBank mRNA AB451282.1 - 2, 3 410
          AB451414.1 - 2, 3 410
          AF009620.1 - 2, 3 410
          AF380342.1 - 3, 4 495
          AF422927.1 - 4, 5 633
          AF422928.1 - 4, 5 607
          AF422929.1 - 3, 4 675
          AK310630.1 - 4, 5 686
          AY429525.1 - 2, 3 141
          BC028223.2 - 2, 3 437
          BC068050.1 - 3, 4 500 6421760
          BX395849.2 - 3, 4 499
          CB990598.1 - 4, 5 631
          EU168332.1 - 2, 3 410
          JN657192.1 - 2, 3 372
          U58143.1 - 2, 3 410
          U60520.1 - 5, 6 762
          X98172.1 - 3, 4 701
          X98173.1 - 3, 4 482
          X98174.1 - 1, 2 25
          X98175.1 - 1, 2 25
          X98176.1 - 4, 5 599
          X98177.1 - 2, 3 410
          X98178.1 - 2, 3 410

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular component disassembly involved in execution phase of apoptosis
          cell surface receptor signaling pathway
          response to cold
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          natural killer cell activation
          macrophage differentiation
          response to cobalt ion
          response to estradiol
          response to lipopolysaccharide
          response to tumor necrosis factor
          TRIF-dependent toll-like receptor signaling pathway
          TRAIL-activated apoptotic signaling pathway
          suppression by virus of host cysteine-type endopeptidase activity involved in apoptotic process
          T cell activation
          B cell activation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of proteolysis
          response to antibiotic
          protein heterooligomerization
          proteolysis involved in cellular protein catabolic process
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          nucleotide-binding oligomerization domain containing signaling pathway
          cellular response to mechanical stimulus
          cellular response to organic cyclic compound
          death-inducing signaling complex assembly
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          execution phase of apoptosis
          activation of cysteine-type endopeptidase activity
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          cysteine-type endopeptidase activity
          death receptor binding
          tumor necrosis factor receptor binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          ubiquitin protein ligase binding
          protein complex binding
          death effector domain binding
          identical protein binding
          scaffold protein binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0