Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COL13A1 SIRNA Assays ›
          Gene Symbol:
          COL13A1
          Gene Name:
          collagen type XIII alpha 1 chain
          Gene Aliases:
          CMS19, COLXIIIA1
          Chromosome Location:
          Chr.10: 69801885 - 69959148 on Build GRCh38
          UniGene ID:
          Hs.695934
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS141633
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COL13A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130103.1 NP_001123575.1 1, 2 835
          NM_001320951.1 NP_001307880.1 1, 2 835
          NM_080798.3 NP_542988.3 1, 2 835
          NM_080800.3 NP_542990.3 1, 2 835
          NM_080801.3 NP_542991.3 1, 2 835
          NM_080802.3 NP_542992.3 1, 2 835
          NM_080805.3 NP_542995.3 1, 2 835
          XM_011539292.2 XP_011537594.1 1, 2 838
          XM_011539293.2 XP_011537595.1 1, 2 838
          XM_011539294.2 XP_011537596.1 1, 2 838
          XM_011539295.2 XP_011537597.1 1, 2 838
          XM_017015676.1 XP_016871165.1 1, 2 838
          XM_017015677.1 XP_016871166.1 1, 2 838
          XM_017015678.1 XP_016871167.1 1, 2 838
          XM_017015679.1 XP_016871168.1 1, 2 838
          XM_017015680.1 XP_016871169.1 1, 2 838
          XM_017015681.1 XP_016871170.1 1, 2 838
          XM_017015682.1 XP_016871171.1 1, 2 838
          XM_017015683.1 XP_016871172.1 1, 2 838
          XM_017015684.1 XP_016871173.1 1, 2 838
          XM_017015685.1 XP_016871174.1 1, 2 838
          XM_017015686.1 XP_016871175.1 1, 2 838
          XM_017015687.1 XP_016871176.1 1, 2 838
          XM_017015688.1 XP_016871177.1 1, 2 838
          XM_017015689.1 XP_016871178.1 1, 2 838
          XM_017015690.1 XP_016871179.1 1, 2 838
          XM_017015691.1 XP_016871180.1 1, 2 838
          XM_017015692.1 XP_016871181.1 1, 2 838
          XM_017015693.1 XP_016871182.1 1, 2 838
          XM_017015694.1 XP_016871183.1 1, 2 838
          XM_017015695.1 XP_016871184.1 1, 2 838
          XM_017015697.1 XP_016871186.1 1, 2 216
          GenBank mRNA AJ293624.1 - 1, 2 796
          BC136385.1 - 1, 2 605 9020372
          M59217.1 - 1, 2 171

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of a branching structure
          endochondral ossification
          cell-matrix adhesion
          single organismal cell-cell adhesion
          cell differentiation
          extracellular matrix organization
          collagen catabolic process
          protein binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wrkvw:80/100.66.78.186:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0