Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DHPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206974.1 | NP_001193903.1 | 2 | 286 | ||
NM_001930.3 | NP_001921.1 | 2 | 435 | |||
NM_013406.2 | NP_037538.1 | 2 | 435 | |||
NR_038192.1 | 2 | 435 | ||||
XM_011527770.1 | XP_011526072.1 | 2 | 435 | |||
XM_011527771.2 | XP_011526073.1 | 2 | 435 | |||
GenBank mRNA | AK291553.1 | - | 2 | 416 | ||
AK304780.1 | - | 2 | 416 | |||
AL520040.3 | - | 2 | 211 | |||
AY336090.1 | - | 2 | 324 | |||
BC000333.2 | - | 2 | 382 | 2822554 | ||
BC014016.2 | - | 2 | 397 | 4129051 | ||
BU844677.1 | - | 2 | 414 | |||
DA524549.1 | - | 2 | 436 | |||
L39068.1 | - | 2 | 351 | |||
U40579.1 | - | 2 | 397 | |||
U79262.1 | - | 2 | 409 |