Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMOX1 SIRNA Assays ›
          Gene Symbol:
          HMOX1
          Gene Name:
          heme oxygenase 1
          Gene Aliases:
          HMOX1D, HO-1, HSP32, bK286B10
          Chromosome Location:
          Chr.22: 35381067 - 35394214 on Build GRCh38
          UniGene ID:
          Hs.517581
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142462
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HMOX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002133.2 NP_002124.1 2 199
          GenBank mRNA AK313120.1 - 2 169
          BC001491.2 - 2 158 3504480
          BP288767.1 - 2 199
          BT019785.1 - 1 92
          CR456505.1 - 2 207
          CR541945.1 - 2 92
          CR541968.1 - 2 92
          GU187011.1 - 3 216
          GU187012.1 - 2 829
          JF323038.1 - 2 96
          M23041.1 - 2 124
          X06985.1 - 2 172

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          endothelial cell proliferation
          wound healing involved in inflammatory response
          negative regulation of leukocyte migration
          heme oxidation
          cellular iron ion homeostasis
          response to oxidative stress
          small GTPase mediated signal transduction
          excretion
          regulation of blood pressure
          cell death
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of muscle cell apoptotic process
          smooth muscle hyperplasia
          cellular response to nutrient
          negative regulation of mast cell cytokine production
          erythrocyte homeostasis
          low-density lipoprotein particle clearance
          regulation of transcription from RNA polymerase II promoter in response to iron
          cellular response to heat
          response to nicotine
          intracellular signal transduction
          heme catabolic process
          response to hydrogen peroxide
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of mast cell degranulation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of neuron apoptotic process
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          response to estrogen
          positive regulation of chemokine biosynthetic process
          regulation of angiogenesis
          positive regulation of angiogenesis
          positive regulation of vasodilation
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          regulation of sequence-specific DNA binding transcription factor activity
          protein homooligomerization
          iron ion homeostasis
          cellular response to arsenic-containing substance
          cellular response to cadmium ion
          cellular response to hypoxia
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          heme oxygenase (decyclizing) activity
          phospholipase D activity
          signal transducer activity
          protein binding
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0