Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NF2 SIRNA Assays ›
          Gene Symbol:
          NF2
          Gene Name:
          neurofibromin 2
          Gene Aliases:
          ACN, BANF, SCH
          Chromosome Location:
          Chr.22: 29603556 - 29698600 on Build GRCh38
          UniGene ID:
          Hs.187898
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS143099
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000268.3 NP_000259.1 1 535
          NM_016418.5 NP_057502.2 1 535
          NM_181825.2 NP_861546.1 1 535
          NM_181828.2 NP_861966.1 1 535
          NM_181829.2 NP_861967.1 1 535
          NM_181830.2 NP_861968.1 1 535
          NM_181831.2 NP_861969.1 1 535
          NM_181832.2 NP_861970.1 1 535
          NM_181833.2 NP_861971.1 1 535
          GenBank mRNA AF122827.1 - 1 142
          AF122828.1 - 1 142
          AF123570.1 - 1 142
          AF369657.1 - 1 517
          AF369658.1 - 1 517
          AF369661.1 - 1 92
          AF369662.1 - 1 92
          AF369663.1 - 1 92
          AF369664.1 - 1 92
          AF369665.1 - 1 92
          AF369666.1 - 1 92
          AF369667.1 - 1 92
          AF369668.1 - 1 92
          AF369701.1 - 1 458
          AK297116.1 - 1 474
          BC003112.1 - 1 364 3537045
          BC020257.1 - 1 430 4871980
          CF138178.1 - 1 316
          CN419318.1 - 1 535
          CR456530.1 - 1 154
          L11353.1 - 1 311
          Z22664.1 - 1 266

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001252251, NM_001252252, NM_001252250, NM_010898, L27105, and others.

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          negative regulation of cell-matrix adhesion
          negative regulation of protein kinase activity
          ectoderm development
          negative regulation of DNA replication
          negative regulation of cell proliferation
          Schwann cell proliferation
          regulation of gliogenesis
          hippocampus development
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          negative regulation of cell growth
          negative regulation of cell migration
          regulation of protein stability
          regulation of hippo signaling
          odontogenesis of dentin-containing tooth
          negative regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of MAPK cascade
          cell-cell junction organization
          positive regulation of cell differentiation
          negative regulation of JAK-STAT cascade
          positive regulation of stress fiber assembly
          lens fiber cell differentiation
          regulation of stem cell proliferation
          regulation of protein localization to nucleus
          regulation of neural precursor cell proliferation
          actin binding
          integrin binding
          protein binding
          beta-catenin binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0