Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NTF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006179.4 | NP_006170.1 | 2 | 629 | ||
XM_005258962.3 | XP_005259019.1 | 3 | 750 | |||
XM_006723232.3 | XP_006723295.1 | 3 | 792 | |||
XM_011527008.2 | XP_011525310.1 | 2 | 533 | |||
XM_011527009.2 | XP_011525311.1 | 2 | 1622 | |||
XM_011527010.2 | XP_011525312.1 | 2 | 1740 | |||
GenBank mRNA | BC012421.2 | - | 2 | 629 | 3865300 | |
BT019368.1 | - | 1 | 488 | |||
BT019369.1 | - | 1 | 488 | |||
CR541900.1 | - | 1 | 488 |