Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098620.2 | NP_001092090.1 | 7 | 789 | ||
NM_001128911.1 | NP_001122383.1 | 7 | 789 | |||
NM_001128912.1 | NP_001122384.1 | 7 | 789 | |||
NM_001128913.1 | NP_001122385.1 | 7 | 789 | |||
NM_001128914.1 | NP_001122386.1 | 7 | 789 | |||
NM_005016.5 | NP_005007.2 | 7 | 789 | |||
NM_031989.4 | NP_114366.1 | 7 | 789 | |||
GenBank mRNA | AB188306.1 | - | 7 | 789 | ||
AB208825.1 | - | 6 | 523 | |||
AK023529.1 | - | 2 | 1388 | |||
AK225357.1 | - | 6 | 503 | |||
AK292141.1 | - | 7 | 566 | |||
AK299210.1 | - | 5 | 383 | |||
AK302067.1 | - | 6 | 506 | |||
BC001155.1 | - | 6 | 527 | 3356341 | ||
BC035420.1 | - | 7 | 762 | |||
BC071942.1 | - | 7 | 779 | 6424433 | ||
BC107688.1 | - | 6 | 455 | 5395926 | ||
CX164318.1 | - | 7 | 781 | |||
X78136.1 | - | 6 | 461 |