Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PHF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002636.4 | NP_002627.1 | 4 | 527 | ||
NM_024165.2 | NP_077084.1 | 4 | 527 | |||
NR_027692.1 | 4 | 527 | ||||
XM_011514662.1 | XP_011512964.1 | 4 | 444 | |||
XM_011514663.1 | XP_011512965.1 | 4 | 444 | |||
XM_011514664.1 | XP_011512966.1 | 4 | 444 | |||
XM_011514665.1 | XP_011512967.1 | 4 | 444 | |||
XM_011514666.1 | XP_011512968.1 | 4 | 444 | |||
XM_011514669.1 | XP_011512971.1 | 4 | 444 | |||
XM_011514670.2 | XP_011512972.1 | 4 | 444 | |||
XM_017010939.1 | XP_016866428.1 | 3 | 239 | |||
GenBank mRNA | AF029678.1 | - | 4 | 347 | ||
AF052205.1 | - | 4 | 343 | |||
AK295412.1 | - | 4 | 397 | |||
AK299162.1 | - | 1 | 26 | |||
AK316499.1 | - | 4 | 478 | |||
BC008834.2 | - | 4 | 495 | 4304525 | ||
DA872118.1 | - | 4 | 532 |