Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU5F1 SIRNA Assays ›
          Gene Symbol:
          POU5F1 POU5F1B POU5F1P3 POU5F1P4
          Gene Name:
          POU class 5 homeobox 1
          POU class 5 homeobox 1B
          POU class 5 homeobox 1 pseudogene 3
          POU class 5 homeobox 1 pseudogene 4
          Gene Aliases:
          OCT3, OCT4, OTF-3, OTF3, OTF4, Oct-3, Oct-4
          OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
          OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
          Oct4-pg4, POU5FLC1
          Chromosome Location:
          Chr.6: 31164337 - 31170693 on Build GRCh38
          Chromosome Location:
          Chr.8: 127415612 - 127417210 on Build GRCh38
          Chromosome Location:
          Chr.12: 8133769 - 8134852 on Build GRCh38
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          UniGene ID:
          Hs.249184
          Hs.450254
          Hs.646545
          Hs.632482
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID HSS143403
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POU5F1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001173531.2 NP_001167002.1 4 997
          NM_001285986.1 NP_001272915.1 2 1708
          NM_001285987.1 NP_001272916.1 3 1483
          NM_002701.5 NP_002692.2 4 838
          NM_203289.5 NP_976034.4 3 1483
          GenBank mRNA AB971680.1 - 4 457
          AB971681.1 - 4 493
          BC117435.1 - 4 819 40125986
          BC117437.1 - 4 819 40125988
          DQ486514.1 - 2 1707
          DQ486515.1 - 3 1151
          DQ486516.1 - 4 665
          DQ486517.1 - 4 473
          GU480808.1 - 4 766
          GU480809.1 - 4 766
          GU480810.1 - 4 766
          GU480811.1 - 4 766
          GU480812.1 - 4 766
          GU480813.1 - 4 766
          GU480815.1 - 4 766
          GU480816.1 - 4 766
          GU480817.1 - 4 766
          GU480818.1 - 4 766
          GU480819.1 - 4 766
          GU480820.1 - 4 766
          GU480821.1 - 4 766
          GU480822.1 - 4 766
          GU480823.1 - 4 766
          GU480824.1 - 4 766
          GU480825.1 - 4 766
          GU480826.1 - 4 766
          GU480827.1 - 4 766
          GU480828.1 - 4 766
          GU480829.1 - 4 766
          GU480830.1 - 4 766
          HQ122658.1 - 4 766
          HQ122659.1 - 4 765
          HQ122660.1 - 4 766
          HQ122661.1 - 4 766
          HQ122662.1 - 4 766
          HQ122663.1 - 4 766
          HQ122664.1 - 4 766
          HQ122665.1 - 4 766
          HQ122666.1 - 4 766
          HQ122667.1 - 4 766
          HQ122668.1 - 4 766
          HQ122669.1 - 4 766
          HQ122670.1 - 4 766
          HQ122671.1 - 4 766
          HQ122672.1 - 4 766
          HQ122673.1 - 4 766
          HQ122674.1 - 4 766
          HQ122675.1 - 4 766
          KF880691.1 - 4 1040
          KF880692.1 - 2 1751
          KF880693.1 - 3 1121
          KJ624996.1 - 4 795
          KJ700311.1 - 3 1121
          KM188057.1 - 3 1364
          Z11898.1 - 4 807
          Z11899.1 - 3 581
          Gene Symbol: POU5F1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA GU480844.1 - 1 766
          Gene Symbol: POU5F1P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036440.1 1 764
          GenBank mRNA AF268617.1 - 1 763
          GU480832.1 - 1 764
          GU480837.1 - 1 764
          GU480838.1 - 1 764
          GU480843.1 - 1 764
          GU480846.1 - 1 764
          GU480847.1 - 1 764
          GU480876.1 - 1 764
          GU480877.1 - 1 764
          GU480887.1 - 1 764
          GU480888.1 - 1 765
          GU480891.1 - 1 765
          GU480893.1 - 1 764
          GU480897.1 - 1 764
          GU480901.1 - 1 763
          GU480903.1 - 1 764
          GU480907.1 - 1 764
          GU480909.1 - 1 764
          GU480915.1 - 1 764
          HQ122692.1 - 1 764
          HQ122693.1 - 1 764
          HQ122694.1 - 1 764
          HQ122722.1 - 1 764
          HQ122723.1 - 1 764
          HQ122724.1 - 1 764
          HQ122727.1 - 1 764
          HQ122728.1 - 1 764
          HQ122753.1 - 1 764
          HQ122754.1 - 1 764
          HQ122755.1 - 1 764
          HQ122791.1 - 1 764
          HQ122792.1 - 1 764
          HQ122793.1 - 1 764
          HQ122794.1 - 1 764
          HQ122796.1 - 1 764
          HQ122797.1 - 1 764
          HQ122828.1 - 1 764
          HQ122829.1 - 1 764
          HQ122830.1 - 1 764
          HQ122831.1 - 1 764
          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034180.1 1 766
          GenBank mRNA AF268613.1 - 1 766
          DQ851566.1 - 1 766
          GU480814.1 - 1 767
          GU480831.1 - 1 768
          GU480833.1 - 1 767
          GU480834.1 - 1 767
          GU480836.1 - 1 767
          GU480839.1 - 1 767
          GU480848.1 - 1 767
          GU480852.1 - 1 767
          GU480853.1 - 1 768
          GU480854.1 - 1 767
          GU480855.1 - 1 765
          GU480856.1 - 1 767
          GU480857.1 - 1 767
          GU480858.1 - 1 767
          GU480859.1 - 1 767
          GU480860.1 - 1 766
          GU480862.1 - 1 767
          GU480865.1 - 1 768
          GU480866.1 - 1 767
          GU480867.1 - 1 767
          GU480869.1 - 1 767
          GU480870.1 - 1 767
          GU480871.1 - 1 767
          GU480872.1 - 1 767
          GU480873.1 - 1 767
          GU480874.1 - 1 767
          GU480875.1 - 1 767
          GU480879.1 - 1 767
          GU480881.1 - 1 767
          GU480882.1 - 1 767
          GU480890.1 - 1 766
          GU480894.1 - 1 767
          GU480895.1 - 1 767
          GU480898.1 - 1 766
          GU480900.1 - 1 766
          GU480904.1 - 1 767
          GU480914.1 - 1 766
          HQ122676.1 - 1 767
          HQ122677.1 - 1 767
          HQ122678.1 - 1 765
          HQ122679.1 - 1 766
          HQ122681.1 - 1 767
          HQ122682.1 - 1 767
          HQ122683.1 - 1 768
          HQ122684.1 - 1 767
          HQ122686.1 - 1 767
          HQ122687.1 - 1 767
          HQ122688.1 - 1 766
          HQ122689.1 - 1 765
          HQ122690.1 - 1 767
          HQ122691.1 - 1 767
          HQ122695.1 - 1 766
          HQ122696.1 - 1 766
          HQ122697.1 - 1 766
          HQ122698.1 - 1 767
          HQ122699.1 - 1 767
          HQ122700.1 - 1 767
          HQ122701.1 - 1 767
          HQ122702.1 - 1 767
          HQ122703.1 - 1 767
          HQ122704.1 - 1 767
          HQ122705.1 - 1 767
          HQ122706.1 - 1 767
          HQ122707.1 - 1 766
          HQ122708.1 - 1 767
          HQ122709.1 - 1 767
          HQ122731.1 - 1 767
          HQ122732.1 - 1 767
          HQ122733.1 - 1 767
          HQ122734.1 - 1 767
          HQ122735.1 - 1 767
          HQ122736.1 - 1 767
          HQ122737.1 - 1 767
          HQ122738.1 - 1 767
          HQ122739.1 - 1 767
          HQ122740.1 - 1 767
          HQ122741.1 - 1 767
          HQ122742.1 - 1 767
          HQ122743.1 - 1 767
          HQ122744.1 - 1 767
          HQ122745.1 - 1 767
          HQ122746.1 - 1 767
          HQ122747.1 - 1 767
          HQ122748.1 - 1 767
          HQ122756.1 - 1 767
          HQ122757.1 - 1 767
          HQ122758.1 - 1 767
          HQ122759.1 - 1 767
          HQ122760.1 - 1 767
          HQ122761.1 - 1 767
          HQ122762.1 - 1 767
          HQ122763.1 - 1 767
          HQ122764.1 - 1 767
          HQ122765.1 - 1 767
          HQ122766.1 - 1 767
          HQ122767.1 - 1 767
          HQ122768.1 - 1 767
          HQ122769.1 - 1 766
          HQ122770.1 - 1 767
          HQ122771.1 - 1 767
          HQ122772.1 - 1 767
          HQ122773.1 - 1 767
          HQ122774.1 - 1 767
          HQ122775.1 - 1 767
          HQ122776.1 - 1 767
          HQ122777.1 - 1 766
          HQ122778.1 - 1 767
          HQ122779.1 - 1 767
          HQ122780.1 - 1 767
          HQ122798.1 - 1 767
          HQ122799.1 - 1 766
          HQ122800.1 - 1 768
          HQ122801.1 - 1 767
          HQ122802.1 - 1 765
          HQ122803.1 - 1 768
          HQ122804.1 - 1 767
          HQ122805.1 - 1 767
          HQ122806.1 - 1 767
          HQ122807.1 - 1 767
          HQ122808.1 - 1 767
          HQ122809.1 - 1 765
          HQ122810.1 - 1 767
          HQ122811.1 - 1 767
          HQ122812.1 - 1 767
          HQ122813.1 - 1 767
          HQ122814.1 - 1 765
          HQ122815.1 - 1 767
          HQ122832.1 - 1 767
          HQ122833.1 - 1 767
          HQ122834.1 - 1 767
          HQ122835.1 - 1 765
          HQ122836.1 - 1 767
          HQ122837.1 - 1 767
          HQ122838.1 - 1 765
          HQ122839.1 - 1 767
          HQ122840.1 - 1 767
          HQ122858.1 - 1 767
          HQ122859.1 - 1 767
          HQ122860.1 - 1 767
          HQ122861.1 - 1 767
          HQ122862.1 - 1 767
          HQ122888.1 - 1 767
          HQ122889.1 - 1 767
          HQ122890.1 - 1 767
          HQ122904.1 - 1 767
          HQ122905.1 - 1 767
          HQ122906.1 - 1 768
          HQ122907.1 - 1 767
          HQ122908.1 - 1 767
          HQ122909.1 - 1 768
          HQ122910.1 - 1 767
          HQ122911.1 - 1 767
          HQ122912.1 - 1 767
          HQ122913.1 - 1 767
          HQ122914.1 - 1 767
          Z21963.1 - 1 205

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          endodermal cell fate specification
          blastocyst development
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          response to wounding
          anatomical structure morphogenesis
          regulation of asymmetric cell division
          regulation of gene expression
          somatic stem cell population maintenance
          mRNA transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of gene silencing by miRNA
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          ubiquitin protein ligase binding
          miRNA binding
          sequence-specific DNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0