Search
Search

PSG4| Gene Symbol: PSG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276495.1 | NP_001263424.1 | 5 | 1821 | ||
| NM_001316339.1 | NP_001303268.1 | 5 | 1734 | |||
| NM_002780.4 | NP_002771.2 | 6 | 2100 | |||
| NM_213633.2 | NP_998798.1 | 5 | 1821 | |||
| XM_017026996.1 | XP_016882485.1 | 7 | 2162 | |||
| XM_017027000.1 | XP_016882489.1 | 6 | 1887 | |||
| GenBank mRNA | AF289606.1 | - | 6 | 1441 | ||
| AK075149.1 | - | 6 | 1715 | |||
| BC008405.1 | - | 8 | 1959 | 4043484 | ||
| BC063127.1 | - | 6 | 1968 | 4768860 | ||
| M33665.1 | - | 4 | 1241 | |||
| M94891.1 | - | 6 | 1948 | |||
| U18468.1 | - | 6 | 1867 | |||
| U18469.1 | - | 5 | 1610 | |||
| X17097.1 | - | 6 | 1955 | |||