Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002931.3 | NP_002922.2 | 2 | 216 | ||
GenBank mRNA | AK289355.1 | - | 2 | 225 | ||
AK297552.1 | - | 2 | 215 | |||
AK301010.1 | - | 2 | 225 | |||
AK301378.1 | - | 2 | 219 | |||
BC002922.2 | - | 2 | 211 | 3954313 | ||
BC051866.2 | - | 2 | 209 | 6142438 | ||
BI259576.1 | - | 2 | 225 |