Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NKX2-1 SIRNA Assays ›
          Gene Symbol:
          NKX2-1 NKX2-1-AS1
          Gene Name:
          NK2 homeobox 1
          NKX2-1 antisense RNA 1
          Gene Aliases:
          BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1
          -
          Chromosome Location:
          Chr.14: 36516397 - 36520225 on Build GRCh38
          Chromosome Location:
          Chr.14: 36519278 - 36523016 on Build GRCh38
          UniGene ID:
          Hs.94367
          Hs.497090
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS144278
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NKX2-1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079668.2 NP_001073136.1 2 252
          NM_003317.3 NP_003308.1 1 407
          GenBank mRNA BC006221.2 - 2 186 3941576
          BE796718.1 - 2 167
          BT009773.1 - 2 156
          D50740.1 - 3 310
          D50741.1 - 1 127
          U19756.1 - 1 66
          U33749.1 - 1 407
          U43203.1 - 2 238
          X82850.1 - 1 80
          Gene Symbol: NKX2-1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_103710.1 11
          GenBank mRNA DA256249.1 - 11

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013093, XM_006240118, XM_006240117, XM_006240116, AB894119, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009385, NM_001146198, BC057607, BC080868, AB221567.

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neuron migration
          regulation of blood volume by renin-angiotensin
          transcription, DNA-templated
          phospholipid metabolic process
          axon guidance
          brain development
          endoderm development
          locomotory behavior
          feeding behavior
          response to hormone
          positive regulation of gene expression
          negative regulation of epithelial to mesenchymal transition
          globus pallidus development
          hippocampus development
          cerebral cortex cell migration
          forebrain dorsal/ventral pattern formation
          forebrain neuron fate commitment
          cerebral cortex GABAergic interneuron differentiation
          pituitary gland development
          lung development
          negative regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          thyroid gland development
          forebrain development
          developmental induction
          response to lipopolysaccharide
          Leydig cell differentiation
          hyperosmotic salinity response
          positive regulation of circadian rhythm
          response to ethanol
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          development of primary female sexual characteristics
          rhythmic process
          anatomical structure formation involved in morphogenesis
          oligodendrocyte differentiation
          lung saccule development
          epithelial tube branching involved in lung morphogenesis
          Clara cell differentiation
          Type II pneumocyte differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region DNA binding
          core promoter binding
          intronic transcription regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          enzyme binding
          transcription regulatory region DNA binding
          intronic transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sdkpq:80/100.66.77.174:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0