Search
Search

ZNF213| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134655.1 | NP_001128127.1 | 5 | 1044 | ||
| NM_004220.2 | NP_004211.1 | 5 | 1063 | |||
| NR_104432.1 | 5 | 1127 | ||||
| XM_011522652.2 | XP_011520954.1 | 5 | 1031 | |||
| GenBank mRNA | AF017433.1 | - | 6 | 986 | ||
| AK055337.1 | - | 5 | 1024 | |||
| AK289834.1 | - | 5 | 1012 | |||
| AK291789.1 | - | 5 | 976 | |||
| AK297454.1 | - | 3 | 364 | |||
| BC007287.1 | - | 5 | 981 | 3347511 | ||
| KU178402.1 | - | 4 | 616 | |||