Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OGT SIRNA Assays ›
          Gene Symbol:
          OGT
          Gene Name:
          O-linked N-acetylglucosamine (GlcNAc) transferase
          Gene Aliases:
          HINCUT-1, HRNT1, O-GLCNAC
          Chromosome Location:
          Chr.X: 71533062 - 71575897 on Build GRCh38
          UniGene ID:
          Hs.405410
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144758
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OGT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_181672.2 NP_858058.1 19 2754
          NM_181673.2 NP_858059.1 19 2724
          XM_017029907.1 XP_016885396.1 17 2283
          XM_017029908.1 XP_016885397.1 17 5756
          GenBank mRNA AF223393.1 - 15 2391
          AK313530.1 - 15 2670
          AL050366.1 - 14 3981
          AL833085.2 - 19 2734
          BC014434.1 - 19 2655 4865031
          BC038180.1 - 19 2687 5017795
          BX537844.1 - 19 2712
          U77413.1 - 15 2403

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          glucosamine metabolic process
          regulation of glycolytic process
          protein O-linked glycosylation
          apoptotic process
          signal transduction
          response to nutrient
          negative regulation of peptidyl-threonine phosphorylation
          positive regulation of granulocyte differentiation
          forebrain development
          negative regulation of protein ubiquitination
          response to insulin
          cellular response to insulin stimulus
          circadian regulation of gene expression
          negative regulation of peptidyl-serine phosphorylation
          regulation of Rac protein signal transduction
          positive regulation of catalytic activity
          histone H4-K5 acetylation
          histone H4-K8 acetylation
          histone H4-K16 acetylation
          positive regulation of cell size
          positive regulation of proteolysis
          positive regulation of transcription from RNA polymerase II promoter
          regulation of insulin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          intracellular distribution of mitochondria
          positive regulation of histone H3-K4 methylation
          negative regulation of cell death
          positive regulation of histone H3-K27 methylation
          protein homotrimerization
          protein heterotrimerization
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to glucose stimulus
          histone H3-K4 trimethylation
          negative regulation of protein targeting to membrane
          regulation of gluconeogenesis involved in cellular glucose homeostasis
          cellular response to toxic substance
          negative regulation of cellular response to hypoxia
          positive regulation of protein localization to nucleus
          positive regulation of reactive oxygen species biosynthetic process
          protein binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          enzyme activator activity
          transcription factor binding
          acetylglucosaminyltransferase activity
          protein N-acetylglucosaminyltransferase activity
          protein domain specific binding
          peptide binding
          histone acetyltransferase activity (H4-K5 specific)
          histone acetyltransferase activity (H4-K8 specific)
          histone acetyltransferase activity (H4-K16 specific)
          monosaccharide binding
          protein O-GlcNAc transferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0