Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320979.1 | NP_001307908.1 | 2 | 391 | ||
NM_003689.3 | NP_003680.2 | 2 | 391 | |||
GenBank mRNA | AF026947.1 | - | 2 | 359 | ||
AK225267.1 | - | 2 | 362 | |||
AK303131.1 | - | 2 | 327 | |||
BC004111.2 | - | 2 | 318 | 3832140 | ||
BC007352.2 | - | 2 | 366 | |||
BC010852.1 | - | 2 | 367 | 3898226 | ||
BC011586.1 | - | 2 | 318 | 4620484 | ||
BC012171.1 | - | 2 | 366 | 4642811 | ||
BC013996.2 | - | 2 | 322 | 3625337 | ||
BG386786.1 | - | 392 | ||||
BT007347.1 | - | 2 | 282 | |||
BX401279.2 | - | 2 | 339 | |||
EU794591.1 | - | 2 | 391 | |||
Y16675.1 | - | 2 | 353 |