Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003731.2 | NP_003722.2 | 2 | 302 | ||
GenBank mRNA | BC000864.1 | - | 2 | 257 | 3459939 | |
BT006766.1 | - | 2 | 177 | |||
DB466912.1 | - | 2 | 301 | |||
Z96932.1 | - | 2 | 223 |
Gene Symbol: LOC107986084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007618.1 | XP_016863107.1 | 2 | 579 |