Search Thermo Fisher Scientific
Gene Symbol: GINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021067.4 | NP_066545.3 | 4, 5 | 466 | ||
XM_017028162.1 | XP_016883651.1 | 4, 5 | 423 | |||
GenBank mRNA | BC012542.1 | - | 4, 5 | 467 | 4333095 | |
D80008.1 | - | 4, 5 | 422 |