Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEC16A SIRNA Assays ›
Gene Symbol:
SEC16A
Gene Name:
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases:
KIAA0310, SEC16L, p250
Chromosome Location:
Chr.9: 136440096 - 136484552 on Build GRCh38
UniGene ID:
Hs.522500
Species:
Human
siRNA ID
HSS145370
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEC16A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276418.1
NP_001263347.1
2
3360
NM_014866.1
NP_055681.1
3
3350
XM_005266134.2
XP_005266191.1
2
3213
XM_011519246.2
XP_011517548.1
3
3359
XM_011519247.1
XP_011517549.1
2
3216
XM_011519248.2
XP_011517550.1
2
3190
XM_011519249.1
XP_011517551.1
2
3213
XM_011519250.2
XP_011517552.1
2
3213
XM_011519251.1
XP_011517553.1
2
3213
XM_011519252.2
XP_011517554.1
2
3213
XM_011519253.1
XP_011517555.1
2
3213
XM_011519254.1
XP_011517556.1
2
3213
XM_011519255.2
XP_011517557.1
2
3213
XM_011519256.1
XP_011517558.1
2
3213
XM_011519257.1
XP_011517559.1
2
3213
XM_011519258.1
XP_011517560.1
2
3213
XM_011519259.1
XP_011517561.1
2
3213
XM_011519260.2
XP_011517562.1
2
3197
XM_011519261.1
XP_011517563.1
2
3213
XM_011519264.2
XP_011517566.1
2
3213
XM_017015363.1
XP_016870852.1
2
3213
XM_017015364.1
XP_016870853.1
2
3213
GenBank mRNA
AB002308.4
-
3
3350
AB593115.1
-
2
3360
BC125018.1
-
1
1922
DQ903855.1
-
1
2507
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endoplasmic reticulum organization
protein transport
substantia nigra development
COPII vesicle coating
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0